Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United States
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Captan D6 100 µg/mL in Acetone, Dr. Ehrenstorfer, LGC Standards
Captan D6 100 µg/mL in Acetone, Dr. Ehrenstorfer, LGC Standards
Captan D6 100 µg/mL in Acetone, Dr. Ehrenstorfer, LGC Standards
77691-860
:
LGC Standards
New Product
:
Captan D6 100 µg/mL in Acetone, Dr. Ehrenstorfer, LGC Standards
77691-860
:
LGC Standards
:
DRE-XA10960100AC
New Product
:
CAS No.:
1330190-00-5
Description:
Captan D6 100 µg/mL in Acetone
Size:
1 ml
Cat. No.:
77691-860