Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United States
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
PCB-Mix 3 100 µg/mL in Isooctane, Dr. Ehrenstorfer, LGC Standards
PCB-Mix 3 100 µg/mL in Isooctane, Dr. Ehrenstorfer, LGC Standards
PCB-Mix 3 100 µg/mL in Isooctane, Dr. Ehrenstorfer, LGC Standards
Catalog # 77679-214
Supplier:
LGC Standards
New Product
CAS Number:
Expand image
PCB-Mix 3 100 µg/mL in Isooctane, Dr. Ehrenstorfer, LGC Standards
Catalog # 77679-214
Supplier:
LGC Standards
Supplier Number:
DRE-X20030300IO
New Product
CAS Number:
Specifications
Description:
PCB-Mix 3 100 µg/mL in Isooctane
Size:
10 ml
Cat. No.:
77679-214
Specifications
About this item
Product Details
Product Details