Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United States
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
PCB No. 149 10 µg/mL in Isooctane, Dr. Ehrenstorfer, LGC Standards
PCB No. 149 10 µg/mL in Isooctane, Dr. Ehrenstorfer, LGC Standards
PCB No. 149 10 µg/mL in Isooctane, Dr. Ehrenstorfer, LGC Standards
77679-155
:
LGC Standards
New Product
:
PCB No. 149 10 µg/mL in Isooctane, Dr. Ehrenstorfer, LGC Standards
77679-155
:
LGC Standards
:
DRE-L20014900IO
New Product
:
CAS No.:
38380-04-0
Description:
PCB No. 149 10 µg/mL in Isooctane
Size:
10 ml
Formula:
C₁₂H₄Cl₆
Molecular Weight:
360.88 g/mol
Cat. No.:
77679-155