Thiamethoxam 10 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
Catalog # 77679-130
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:153719-23-4
- Description:Thiamethoxam 10 µg/mL in Acetonitrile
- Size:10 ml
- Formula:C₈H₁₀ClN₅O₃S
- Molecular Weight:291.72 g/mol
- Cat. No.:77679-130