PCB No. 151, Dr. Ehrenstorfer, LGC Standards
Catalog # 77678-966
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:52663-63-5
- Description:PCB No. 151
- Size:5 mg
- Formula:C₁₂H₄Cl₆
- Molecular Weight:360.88 g/mol
- Cat. No.:77678-966