Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United States
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Standards
Organic Standards
Acetamiprid D3 (N-methyl D3), Dr. Ehrenstorfer, LGC Standards
Acetamiprid D3 (N-methyl D3), Dr. Ehrenstorfer, LGC Standards
Acetamiprid D3 (N-methyl D3), Dr. Ehrenstorfer, LGC Standards
77677-404
:
LGC Standards
New Product
:
Acetamiprid D3 (N-methyl D3), Dr. Ehrenstorfer, LGC Standards
77677-404
:
LGC Standards
:
DRE-C10013100
New Product
:
CAS No.:
1353869-35-8
Description:
Acetamiprid D3 (N-methyl D3)
Size:
50 mg
Cat. No.:
77677-404