$0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
United States
ContactUsLinkComponent
Standards
Organic Standards
Tolpyralate 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
Tolpyralate 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
Tolpyralate 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
77693-439
:
LGC Standards
New Product
:
Tolpyralate 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
77693-439
:
LGC Standards
:
DRE-A17591900AL-100
New Product
:
CAS No.:
1101132-67-5
Description:
Tolpyralate 100 µg/mL in Acetonitrile
Size:
1 ml
Cat. No.:
77693-439