(-)-Cotinine 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
Catalog # 77691-581
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:486-56-6
- Description:(-)-Cotinine 100 µg/mL in Acetonitrile
- Size:1 ml
- Formula:C₁₀H₁₂N₂O
- Molecular Weight:176.22 g/mol
- Cat. No.:77691-581