Order Entry
United States
ContactUsLinkComponent
Chloroparaffin C18 60% Cl 100 µg/mL in Cyclohexane, Dr. Ehrenstorfer, LGC Standards
Chloroparaffin C18 60% Cl 100 µg/mL in Cyclohexane, Dr. Ehrenstorfer, LGC Standards
Catalog # 77691-559
Supplier:  LGC Standards
New Product
CAS Number:  
undefined
Chloroparaffin C18 60% Cl 100 µg/mL in Cyclohexane, Dr. Ehrenstorfer, LGC Standards
Catalog # 77691-559
Supplier:  LGC Standards
Supplier Number:  DRE-A11457600CY-100
New Product
CAS Number:  

Specifications

  • CAS No.:
    85535-86-0
  • Description:
    Chloroparaffin C18 60% Cl 100 µg/mL in Cyclohexane
  • Size:
    1 ml
  • Cat. No.:
    77691-559

Specifications

About this item