Albendazole-2-aminosulfone 100 µg/mL in Acetonitrile, Dr. Ehrenstorfer, LGC Standards
Catalog # 77691-448
Supplier: LGC Standards
New Product
CAS Number:
Specifications
- CAS No.:80983-34-2
- Description:Albendazole-2-aminosulfone 100 µg/mL in Acetonitrile
- Size:1 ml
- Formula:C₁₀H₁₃N₃O₂S
- Molecular Weight:239.30 g/mol
- Cat. No.:77691-448