Order Entry
United States
ContactUsLinkComponent
Azido PEG Biotin ≥95%
Azido PEG Biotin ≥95%
Catalog # ALA163284100MG
Supplier:  ALADDIN SCIENTIFIC
New Product
CAS Number:  
Azido PEG Biotin ≥95%
Catalog # ALA163284100MG
Supplier:  ALADDIN SCIENTIFIC
Supplier Number:  A163284-100MG
New Product
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Cat. No.:
    ALA163284100MG

Specifications

About this item

 :
CAS Number:32130-27-1
Formula:NH₂(CH₂CH₂O)nH