Order Entry
United States
ContactUsLinkComponent
Tyrphostin AG 490 ≥98%, Moligand™
Environment
Toxic
Tyrphostin AG 490 ≥98%, Moligand™
Catalog # ALA126525250MG
Supplier:  ALADDIN SCIENTIFIC
New Product
CAS Number:  
undefined
Environment
Toxic
Tyrphostin AG 490 ≥98%, Moligand™
Catalog # ALA126525250MG
Supplier:  ALADDIN SCIENTIFIC
Supplier Number:  A126525-250MG
New Product
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Cat. No.:
    ALA126525250MG

Specifications

About this item

 :
 :294.31 g/mol
Melting Pt:215°C
 :GHS09
 :GHS06
Storage Temperature:Freezer
CAS Number:133550-30-8
 :MFCD00209833
Formula:C₁₇H₁₄N₂O₃
MDL Number:MFCD00236452