Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United States
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Ammonium sulphate
Ammonium sulfate ≥99.99% (metals basis), PrimorTrace™
Ammonium sulfate ≥99.99% (metals basis), PrimorTrace™
Ammonium sulfate ≥99.99% (metals basis), PrimorTrace™
ALA112105-500G
:
ALADDIN SCIENTIFIC
New Product
:
Ammonium sulfate ≥99.99% (metals basis), PrimorTrace™
ALA112105-500G
:
ALADDIN SCIENTIFIC
:
A112105-500G
New Product
:
:
132.14 g/mol
:
>280 °C
:
1.07 g/cm³
:
231-984-1
:
GHS07
:
GHS09
:
7783-20-2
:
:
(NH₄)₂SO₄
:
MFCD00003391