PROPOR MR DEMI Capsules
Supplier: PARKER HANNIFIN TE
New Product
Certificates
About this item
PROPOR MR filters have been specifically designed for the fast, effective, and economical removal of mycoplasma from cell culture media in the biopharmaceutical industry.
- Fully validated and integrity testable for assurance of sterility
- A typical LRV of >10 for Acholeplasma laidlawii for effective mycoplasma control
- Integral prefilter layer increases throughputs for reduction of filter trains and exceptional flow rates for quick processing of cell culture media
Incorporating a highly retentive 0.1 µm rated PES membrane, PROPOR MR is validated against the industry standard Brevundimonas diminuta as well as Acholeplasma laidlawii, a common Mycoplasma species found in contaminated cell cultures.
An asymmetric integral membrane prefilter layer provides PROPOR MR with the optimal membrane configuration for maximum capacity and flow rate. Quick processing times minimize the risk of contamination while still offering maximum protection from Mycoplasma.
PFAS-free options - not with Viton seals.
Ordering information: Downstream filling bell available for 0.25" HB. Supplied with protection cups on vent and drain ports.
Packaging: DEMI capsules are supplied 3 per pack.
Frequently Bought Together