Order Entry
United States
ContactUsLinkComponent
Light Source Assay, FO-6000
Light Source Assay, FO-6000
Catalog # 77664-340
New Product
Light Source Assay, FO-6000
Catalog # 77664-340
Supplier Number:  FO-6000
New Product

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    FO-6000 light source assay
  • Cat. No.:
    77664-340

Specifications

About this item

The FO-6000 is a continuous fiber optic light source featuring an extended visible part of the light source (380 to 1700 nm). It has a SMA fiber optic connector. Both, shutter and lamp can be controlled via a switch or external TTL triggering. This light source offers a wide assortment of applications. A special feature of the FO-6000 is its color balancing optics, which shifts the usable range of the light source from traditional 420 nm down to 380 nm wavelength. Due to its thermally controlled optical bench, it is particularly suitable for low noise and low drift applications.

  • Visible light source and high light output
  • 380 to 1700 nm
  • Low drift <0.5 mAU/h
  • SMA fiber connection
  • Electrical shutter control (switch and TTL)
  • Compact - lightweight and ow power consumption
  • External triggering
  • 15 to 20 minutes warming up time
  • Negligible heat generation

Benefits:
Bulb with 10000 h lifetime
Fits all of WPI's fiber optic probes and flow cells
Design matched to LWCC applications
Temperature controlled optical bench

Applications:
Low noise VIS absorbance measurements
Trace analysis of nutrients in seawater and freshwater
CDOM analysis in seawater
Analytical chemistry, environmental science and life science