Order Entry
United States
ContactUsLinkComponent
Human Recombinant PDGF-AA (from E. coli)
Human Recombinant PDGF-AA (from E. coli)
New Product
Certificates
Human Recombinant PDGF-AA (from E. coli)
New Product

Specifications

  • Conjugation:
    Unconjugated
  • Protein Function:
    Cytokine
  • Protein/Peptide Type:
    Recombinant
  • Source:
    E. coli
  • Species:
    Human
  • Storage Conditions:
    At or below –20 °C
  • Endotoxin Content:
    Endotoxin LAL (EU/µg) ≤0.01
  • Biological Activity:
    Bioactivity assay: Proliferation of NR6R-3T3 , ED50 ≤50 ng/ml, Bioactivity ≥WHO standard not available
  • Reconstitution Instructions:
    Reconstituted material should be apportioned in working volumes and stored at or below –20 °C in manual defrost freezer.
  • Endotoxin-free:
    N
  • Carrier-Free:
    Y
  • Protease-free:
    N
  • Animal-Free:
    Y
  • Protein Synonyms:
    ODGF|GDGF
  • Protein/Peptide Name:
    PDGF-AA
  • Purity:
    ≥97%
  • Molecular Weight:
    14.4/28.9 kDa
  • Sequence:
    MSIEEAVPAV CKTRTVIYEI PRSQVDPTSA NFLIWPPCVE VKRCTGCCNT SSVKCQPSRV HHRSVKVAKV EYVRKKPKLK EVQVRLEEHL ECACATTSLN PDYREEDTGR PRESGKKRKR KRLKPT
  • Endotoxin Level:
    Low
  • Formulation:
    0.1% Trifluoroacetic Acid (TFA)
  • Nuclease-free:
    N
  • Grade:
    CTGrade GMP
  • Shipping Temperature:
    Ambient
  • Tested Applications:
    Bioassay

Specifications

Related Information

Frequently Bought Together