Order Entry
United States
ContactUsLinkComponent
TCLP Base/Neutral/Acid Extractable Spike Solution
TCLP Base/Neutral/Acid Extractable Spike Solution
Catalog # SXTCLP-BNA
Supplier:  SPEX CERTIPREP LLC
New Product
CAS Number:  
undefined
TCLP Base/Neutral/Acid Extractable Spike Solution
Catalog # SXTCLP-BNA
Supplier:  SPEX CERTIPREP LLC
Supplier Number:  TCLP-BNA
New Product
CAS Number:  

Specifications

  • Cat. No.:
    SXTCLP-BNA
  • Storage temperature:
    Ambient

Specifications

About this item

Semivolatiles US EPA method 1311-TCLP series methods - EPA methods 1311 and 1312 analytical methods for the monitoring of a wide range of compounds in various samples using the Toxicity Characteristic Leaching Procedure (TCLP).