Surrogate Standard
Product Details & Documents
Volatiles, US EPA method 524.3 is a GC/MS method for the analysis of purgeable VOCs in finished drinking water. Analytes added to Methanol 524.3 revision 4 are supplied in a 24-component mix.
Specifications
Specifications
- Storage temperature:Ambient