Orders LinkContactUsLinkComponent
Internal Volatile Standard
Internal Volatile Standard
Ambient
  SX5242-I
Supplier #:  5242-I
Internal Volatile Standard
  SX5242-I
 :  
Internal Volatile Standard
Internal Volatile Standard
Ambient
 :  
$48.73
Each(1/Itm/Bx)
Volatiles, US EPA method 524.3 is a GC/MS method for the analysis of purgeable VOCs in finished drinking water. Analytes added to Methanol 524.3 revision 4 are supplied in a 24-component mix.
  • Storage temperature:
    Ambient