Order Entry
United States
ContactUsLinkComponent
Swiss Microdissection Tweezers, Curved
Swiss Microdissection Tweezers, Curved
Catalog # 77663-644
New Product
Swiss Microdissection Tweezers, Curved
Catalog # 77663-644
Supplier Number:  504505
New Product
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Swiss made microdissection tweezers
  • Length:
    11.5 cm (4.5")
  • Tip configuration:
    Curved
  • Tip dimensions:
    0.18×0.2 mm
  • Cat. No.:
    77663-644

Specifications

About this item

Swiss microdissecting forceps have curved fine tips for precision work in microsurgery, microdissection, and applications requiring fine manipulation of tissue or small objects under a microscope.

  • Acid-resistant/antimagnetic
  • Surgical grade stainless steel

These are hand forceps with a spring action letting you pinch the blades together to control the amount of pressure used. They are manufactured from an acid-resistant, anti-magnetic alloy, and they works well in an MRI environment. These surgical forceps are quality Swiss-made instruments. They may be sterilize using an autoclave or by chemical sterilization.

Low carbon austenitic steel. Contains from 16.5 to 18.5 by weight of chromium and has important quantities of nickel and molybdenum as additional alloying elements. Non-magnetizable, good corrosion resistance to most chemicals, salts and acids.

Typical applications include tweezers for the electronic industry, watch-makers, jewelers and laboratory and medical applications in moderately aggressive chemical environments.