About this item
Horizontal airflow is created using a turbo blower that re-circulates air over the heating element, providing fast heat-up and recovery times, and ensuring a more rapid and efficient rate of drying.
- True horizontal airflow design
- Programmable 24-step ramp and soak controller
- Type-304 stainless steel interior and exterior
- Durable powder-coat paint exterior finish
- Three-inch intake and exhaust ports
- Equipped with adjustable stainless steel shelves
- VWR® Two-Year Limited Parts and Labor Warranty
By forcing continuously warmed air into the chamber, the turbo blower assures uniform heating across all shelves and to every area of the chamber, eliminating concerns of obstructed air, regardless of how the shelves are loaded.
To maintain temperature uniformity and stop heat loss, a 8.9 cm (3¹/₂") thick, double-layer insulating barrier of high-density alumina silicate and high-temperature fiberglass wraps around all five sides of the chamber. A high temperature silicone rubber door seal eliminates air leakage. In addition, the door latch and hinges are user-adjustable to maintain a better seal as metal expands and contracts over the life of the oven.
The independent over-temperature safety is user-adjustable and protects the unit against damage to contents and burnout in the unlikely event that the main controller fails. The sealed membrane touch-pad control panel guards against any harm to the electronics.
Stainless steel interior and exterior provide maximum corrosion resistance. Exterior dimensions include handle and exhaust port.
Specifications
- Catalog No:
- 52202-258
- 52202-509
- Temperature Range:
- 15 °C above ambient to 260 °C
- 15 °C above ambient to 260 °C