Orders LinkContactUsLinkComponent
Cuvettes
Cuvettes
Supplier:   SARSTEDT INC
Cuvettes
Supplier:  SARSTEDT INC
Certificates
Cuvettes
3 Options Available Below
Product Details & Documents
SARSTEDT has been known as a manufacturer of high-quality disposable cuvettes.
  • Height: 45 mm
  • Optical sides: 2
  • Path length: 10 mm
  • Disposable cuvette made of high-quality, crystal-clear polystyrene or acrylic (PMMA)
  • Ideal for photometric analyses of liquids in the wavelength range ≥300 nm (PMMA)/≥330 nm (PS)
  • An arrow integrated in the cuvette allows exact positioning in the photometer
  • Nest sorting ensures consistent transmission values within a polystyrene box
SARSTEDT cuvettes are made from polystyrene (PS), acrylic (PMMA) and a special plastic for analysis in the UV range, which are ideal for photometric analysis of liquid solutions. The cuvettes are also available in 2× optical semi-micro and a 4× optical format for 90° angle fluorescence measurements.
Product Family Options
Product Information
  • Packaging
    Material
    Availability
    Price
  • 77588-356
    Cuvettes
    Cuvettes
    Rack pack
    PMMA
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $28.07
    $28.07
    Log in for availability
    Case of 2000
    Case of 2000 - $561.40
    $561.40
    Pack of 100
       $28.07
    Case of 2000
       $561.40
    Supplier Number: 67.738
  • 77588-360
    Cuvettes
    Cuvettes
    Rack pack
    PS
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 100
    Pack of 100 - $18.53
    $18.53
    Log in for availability
    Case of 2000
    Case of 2000 - $370.60
    $370.60
    Pack of 100
       $18.53
    Case of 2000
       $370.60
    Supplier Number: 67.741
  • 77588-366
    Cuvettes
    Cuvettes
    BAGA
    PS
    Log in for availability
    Log in for availability
    Log in for availability
    Pack of 500
    Pack of 500 - $81.57
    $81.57
    Log in for availability
    Case of 2000
    Case of 2000 - $326.28
    $326.28
    Pack of 500
       $81.57
    Case of 2000
       $326.28
    Supplier Number: 67.745