Tubes with Two-position Closure, 5 and 13 ml
Supplier: SARSTEDT INC
New Product
Certificates
About this item
To keep cells happy, the selection of a suitable cultivation surface is essential. SARSTEDT offers a wide range of cell culture labware and accessories for many applications.
- Purity: Sterile
- Closure: Ventilation cap
- Base shape: Round base
- Tube with round base and 2-position closure
- Made of clear polypropylene or polystyrene to easy inspect the sample material
- Ventilation cap with two-position closure
- First position: Ventilation of the inside of the tube through the only lightly fitted stopper
- Second position: Tube tightly sealed by pushing down the stopper
Tubes with assembled two-position ventilation cap, sterile.
Frequently Bought Together