ContactUsLinkComponent
Felt Pens, Alcohol-resistant
Felt Pens, Alcohol-resistant
Catalog # 77589-994
Supplier:  SARSTEDT INC
New Product
undefined
Felt Pens, Alcohol-resistant
Catalog # 77589-994
Supplier:  SARSTEDT INC
Supplier Number:  95.950
New Product
Specifications
  • Description:
    Felt pens, alcohol-resistant, black ink
  • Environmentally Preferable:
  • Cat. No.:
    77589-994
Specifications
About this item
Hiqh quality labware from SARSTEDT.
  • Permanent labeling of many surfaces used in the laboratory
  • Alcohol-resistant on polystyrene and polycarbonate surfaces (e.g. cell culture dishes)
  • Limited suitability for polypropylene surfaces
  • For dry, wet and cold surfaces
  • Fine tip
Waterproof and fast-drying felt markers from SARSTEDT are perfect for non-smear labeling of plastic tubes and reaction tubes.