Order Entry
United States
Orders LinkContactUsLinkComponent
 

Tissue Culture Inserts, 12-well

Tissue Culture Inserts, 12-well
 
Tissue Culture Inserts, 12-well
Tissue Culture Inserts, 12-well

 

 

To keep cells happy, the selection of a suitable cultivation surface is essential. SARSTEDT offers a wide range of cell culture labware and accessories for many applications.

  • Suitable for: 12-well plates
  • Membrane material: PET
  • Frame material: PS
  • Growth area: 1.1 cm²
  • Working volume, insert: 0.2 to 0.8 ml
  • Working volume, well: 1.2 to 2.4 ml
  • Purity: Sterile, non-pyrogenic/endotoxin-free, non-cytotoxic
  • Inserts for cell culture plates form a 2-compartment system to simulate the in vivo situation in complex experiments in cell and tissue culture
  • Surface treatment on both sides (TC treatment) for better cell adhesion
  • High chemical resistance ensures that cells can be easily fixated and stained and minimizes non-specific binding of molecules
  • User-friendly design: stable, transparent casing, asymmetric for easy pipetting, lowered top edge for optimal gas exchange

Tissue culture (TC) inserts are easy-to-use inserts for TC plates. When used in combination with our plates, the TC inserts form a two-compartment cell culture system in which the in vivo conditions of cells can be simulated extremely well.

Product Family Options

Translucent

 

  • Description
    Color
    Membrane diameter
    Pore density
    Pore size
    Packaging
     
     
  • TC inserts, 12-well
    Translucent
    12 µm
    1×108
    0.4 µm
    Blister pack
       
       
       
    Pack of 48
    Pack of 48 - $390.32
    $390.32
       
    Case of 1440
    Case of 1440 - $11,709.60
    $11,709.60
    Pack of 48
       $390.32

    Case of 1440
       $11,709.60

     : 83.3931.040
  • TC inserts, 12-well
    Translucent
    9 µm
    2×106
    3 µm
    Blister pack
       
       
       
    Pack of 48
    Pack of 48 - $390.32
    $390.32
       
    Case of 1440
    Case of 1440 - $11,709.60
    $11,709.60
    Pack of 48
       $390.32

    Case of 1440
       $11,709.60

     : 83.3931.300
  • TC inserts, 12-well
    Translucent
    10 µm
    6×105
    5 µm
    Blister pack
       
       
       
    Pack of 48
    Pack of 48 - $390.32
    $390.32
       
    Case of 1440
    Case of 1440 - $11,709.60
    $11,709.60
    Pack of 48
       $390.32

    Case of 1440
       $11,709.60

     : 83.3931.500
  • TC inserts, 12-well
    Translucent
    11 µm
    2×105
    8 µm
    Blister pack
       
       
       
    Pack of 48
    Pack of 48 - $390.32
    $390.32
       
    Case of 1440
    Case of 1440 - $11,709.60
    $11,709.60
    Pack of 48
       $390.32

    Case of 1440
       $11,709.60

     : 83.3931.800

Transparent

 

  • Description
    Color
    Membrane diameter
    Pore density
    Pore size
    Packaging
     
     
  • TC inserts, 12-well
    Transparent
    12 µm
    2×106
    0.4 µm
    Blister pack
       
       
       
    Pack of 48
    Pack of 48 - $390.32
    $390.32
       
    Case of 1440
    Case of 1440 - $11,709.60
    $11,709.60
    Pack of 48
       $390.32

    Case of 1440
       $11,709.60

     : 83.3931.041
  • TC inserts, 12-well
    Transparent
    11 µm
    2×106
    1 µm
    Blister pack
       
       
       
    Pack of 48
    Pack of 48 - $390.32
    $390.32
       
    Case of 1440
    Case of 1440 - $11,709.60
    $11,709.60
    Pack of 48
       $390.32

    Case of 1440
       $11,709.60

     : 83.3931.101