Specifications
- Description:Leica MZ10F for fluorescence sorting
- Magnification:8 - 80× (1.0× objective, 10× eyepieces)
- Cat. No.:77676-396
Specifications
About this item
The Leica MZ10-F stereo microscope is a state-of-the-art instrument for precise microscopic observation. Its advanced optics and ergonomic design make it a smart choice for sorting. With exceptional image quality and versatility, the MZ10-F improves efficiency and accuracy in life sciences research.
- 10:1 zoom range with high numerical aperture of 0.125 for excellent representation of small structures
- 8× to 80× standard magnification zoom: For routine sorting tasks for convenient observation from overview to detail
- TripleBeam™, Leica's patented separate beam path for extremely intense fluorescence illumination - variety of standard and custom filters available for specific fluorescence applications
- Fast interchange: FLUOIII™, Leica's patented, four-position, rapid change filter system - allows fast interchange between 4 different filter sets for easy, convenient imaging
The Leica MZ10-F stereo microscope is a cutting-edge instrument designed for precision and clarity in microscopic observation. With its advanced optics and ergonomic design, this microscope is particularly well-suited for cell sorting applications. Delivering exceptional image quality and versatility, the Leica MZ10-F enhances the efficiency and accuracy of cell sorting processes, making it an indispensable tool for researchers and professionals in the field of life sciences.
Features and specifications includes:
Leica MZ10 F optics carrier with zoom magnification changer 10:1
Click stop zoom settings
Triple Beam TM: Separate illumination beam path for fluorescence
Four position fluorescence filter turret
Objective thread M65