Order Entry
United States
ContactUsLinkComponent
Digital Microscope for Easy Confluency Checks, Mateo TL, Leica Microsystems
Digital Microscope for Easy Confluency Checks, Mateo TL, Leica Microsystems
Catalog # 77676-388
Supplier:  Leica Microsystems
New Product
undefined
Digital Microscope for Easy Confluency Checks, Mateo TL, Leica Microsystems
Catalog # 77676-388
Supplier:  Leica Microsystems
Supplier Number:  8122659
New Product

Specifications

  • Description:
    Leica Mateo TL for easy confluency checks
  • Accessories Included:
    Wireless mouse, mouse pad, dust protection cover, power adapter, power cord, quick start guide and user manual
  • Camera:
    Megapixel color camera
  • Condenser:
    S40/0.45
  • Display:
    15.6", 1920×1080 monitor
  • Illumination:
    LED
  • Stage:
    Fixed stage (L×W) 262×212 mm
  • Cat. No.:
    77676-388

Specifications

About this item

The Leica Mateo TL is a digital microscope designed for cell culture checks. With an integrated camera, advanced optics, and automatic confluency evaluations, it streamlines workflow in your lab for seamless and efficient confluency determination.

  • Get started right away with a ready-to-use system
  • Go from setup to first image in under one minute
  • Wirelessly transfer images directly to your smartphone or tablet
  • Get consistent confluency measurements with one common tool (requires software license)

The Leica Mateo TL for easy confluency checks is an innovative digital microscope tailored for routine checks and documentation of cell cultures. Featuring an integrated camera, advanced optics, and proven algorithms, this microscope ensures quick and accurate automatic confluency evaluations. Streamline your workflow with the Leica Mateo TL, making confluency determination a seamless and efficient task in your cell culture lab.

Features and specifications include:
Two objectives are pre-installed: HI PLAN 4×/0.10 PH0, HI PLAN I 10×/0.22 PH1
Three position slider for S40 condenser (BF, PH0, PH1)
Four position objective nosepiece
1× USB 3.0 port and 2× USB 2.0 ports

Delivery: Includes Mateo TL IVD; object guide kit; wifi dongle; 20× HI PLAN objective; confluency module; US power cord.