Visual Comparator Colour Kits, Phenols
104086-334
: Hach
:
- Chemical method:4-Aminoantipyrine
- Description:Phenols test kit, model PL-1
- Cat. No.:104086-334
- Range:0 - 4 mg/L
Simple, economical, rapid visual color determination.
: Kit includes four reagents, two tubes, two color discs, color comparator box, instruction sheet, and carrying case.