ContactUsLinkComponent
Visual Colour Kits, Swimming Pools, Pool Master
Visual Colour Kits, Swimming Pools, Pool Master
Catalog # 104086-100
Supplier:  Hach
CAS Number:  
undefined
Visual Colour Kits, Swimming Pools, Pool Master
Catalog # 104086-100
Supplier:  Hach
Supplier Number:  2315001
CAS Number:  
Specifications
  • Description:
    Pool master test kit
  • Cat. No.:
    104086-100
Specifications
About this item
Pool Master test kit combines several tests into a single test kit. The test kit includes tests for alkalinity (20 to 400 mg/L), free and total chlorine (0 to 3.4 mg/L), hardness (20 to 400 mg/L), and pH (6.6 to 8.4 mg/L) and specialized tests for total alkalinity, calcium hardness, and cyanuric acid.
Delivery: Kit includes ten reagents, clippers, graduated cylinder, sample cell with cap, dipstick, two color discs, viewing tubes, comparator box, instructions and carrying case.