Orders LinkContactUsLinkComponent
Visual Colour Kits, Phosphate
Visual Colour Kits, Phosphate
Total phosphate test kit, model PO-24, 0 - 0.8 mg/L; 0 - 4 mg/L; 0 - 40 mg/L
Catalog # 104086-004
Marketsource
Supplier:   Hach
Supplier #:  225001
Visual Colour Kits, Phosphate
Catalog # 104086-004
Supplier:  Hach
Marketsource
CAS Number:  
Visual Colour Kits, Phosphate
Visual Colour Kits, Phosphate
Total phosphate test kit, model PO-24, 0 - 0.8 mg/L; 0 - 4 mg/L; 0 - 40 mg/L
CAS Number:  
Price
$459.69
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
This kit provides measurement of ortho- (reactive) and meta- (condensed) phosphate (requires hot acid digestion).
Delivery information: Kit includes five reagents, color disc, comparator box, viewing adapter, two viewing tubes, bottle, flask, dropper tube, stove and cover, test tube clamp, funnel, filter paper, deionized water, instructions and carrying case.
Specifications
Specifications
  • Description:
    Total phosphate test kit, model PO-24
  • Range:
    0 - 0.8 mg/L; 0 - 4 mg/L; 0 - 40 mg/L