Order Entry
United States
ContactUsLinkComponent
Visual Comparator Test Kits, Nitrate, NI-11
Visual Comparator Test Kits, Nitrate, NI-11
Catalog # 104085-526
Supplier:  Hach
CAS Number:  
undefined
Visual Comparator Test Kits, Nitrate, NI-11
Catalog # 104085-526
Supplier:  Hach
Supplier Number:  146803
CAS Number:  

Specifications

  • Description:
    Color disc, model NI-11
  • Cat. No.:
    104085-526
  • Range:
    0 - 40 mg/L
  • Type:
    Cadmium reduction

Specifications

About this item

Simple, low cost, portable, color disc method of testing nitrate.

Delivery: Kit includes color disc, comparator box and tubes, reagents, instructions and carrying case.