Visual Colour Comparator Kit, Phosphate
104086-002
: Hach
:
- Description:Phosphate test kit, (ortho+meta), model PO-23
- Cat. No.:104086-002
- Range:0.1 - 4 mg/L; 1 - 40 mg/L
Simple, economical rapid color disc method with an easy to carry test kit.
: Kit includes three reagents, color disc, comparator box, two viewing tubes, bottle, dropper tube, stove and cover, test tube clamp, deionized water, instructions and carrying case.