Chromium Test Kits, Comparator Method, Models CH-8 and CH-12
Catalog # 104085-946
Supplier: Hach
CAS Number:
Specifications
- Chemical method:Diphenylcarbazide hypobromite oxidation
- Description:Chromium test kit, model CH-12, color disc
- Cat. No.:104085-946
- Range:0 - 1.3 mg/L
Specifications
About this item
These kits are used to determine chromium or chromium(VI) using simple, economical color disc methods.
Delivery: CH-12 kit includes four reagents, clippers, two colour discs, comparator box, two viewing tubes, bottle, flask, dropper tube, stove and cover, test tube clamp, instructions and carrying case. CH-8 kit includes one reagent, two colour discs, comparator box, two viewing tubes, vial, instructions and carrying case.