Order Entry
United States
Orders LinkContactUsLinkComponent
 

Buffer Powder Pillows

Marketsource
Supplier:   Hach
Buffer Powder Pillows
Supplier:  Hach
Marketsource
Certificates
Buffer Powder Pillows
Buffer Powder Pillows

13 Options Available Below

Product Details & Documents

The unit-dose powder pillows assure buffer freshness and eliminate the possibility of contamination. Each powder pillow prepares 50 ml of buffer solutions of respective pH.

  • Fresh and reliable
  • No danger of contamination
  • Traceable to NIST standards

 : The buffer pillows are delivered in an airtight container and are supplied with a certificate of conformity.

Product Family Options

Product Information

  • pH accuracy
    pH buffer type
    Reference temperature
    Packed
    pH
    Color
    Availability
    Price
  • ±0.02
    Phosphate
    25 °C
    Pack of 15
    8
    Colorless
    Log in for availability
    Log in for availability
    Pack of 15
    Pack of 15 - $31.28
    $31.28
    Pack of 15
       $31.28
     

    Supplier Number: 1407995
    Specifications:
    Marketsource
    Marketsource
  • ±0.02
    Phosphate
    25 °C
    Pack of 15
    6.86
    Colorless
    Log in for availability
    Log in for availability
    Pack of 15
    Pack of 15 - $31.28
    $31.28
    Pack of 15
       $31.28
     

    Supplier Number: 1409895
    Specifications:
    Marketsource
    Marketsource
  • ±0.02
    Borate
    25 °C
    Pack of 50
    9
    Colorless
    Log in for availability
    Log in for availability
    Pack of 50
    Pack of 50 - $54.20
    $54.20
    Pack of 50
       $54.20
     

    Supplier Number: 1410766
    Specifications:
    Marketsource
    Marketsource
  • ±0.02
    Carbonate
    25 °C
    Pack of 50
    10.01
    Blue
    Log in for availability
    Log in for availability
    Pack of 50
    Pack of 50 - $48.80
    $48.80
    Pack of 50
       $48.80
     

    Supplier Number: 2227166
    Specifications:
    Marketsource
    Marketsource
  • ±0.02
    Carbonate
    25 °C
    Pack of 15
    10.01
    Blue
    Log in for availability
    Log in for availability
    Pack of 15
    Pack of 15 - $26.09
    $26.09
    Pack of 15
       $26.09
     

    Supplier Number: 2227195
    Specifications:
    Marketsource
    Marketsource
  • ±0.02
    Phthalate
    25 °C
    Pack of 250
    4.01
    Red
    Log in for availability
    Log in for availability
    Pack of 250
    Pack of 250 - $200.77
    $200.77
    Pack of 250
       $200.77
     

    Supplier Number: 2226964
    Specifications:
    Marketsource
    Marketsource
  • ±0.02
    Phthalate
    25 °C
    Pack of 50
    4.01
    Red
    Log in for availability
    Log in for availability
    Pack of 50
    Pack of 50 - $48.80
    $48.80
    Pack of 50
       $48.80
     

    Supplier Number: 2226966
    Specifications:
    Marketsource
    Marketsource
  • ±0.02
    Phthalate
    25 °C
    Pack of 15
    4.01
    Red
    Log in for availability
    Log in for availability
    Pack of 15
    Pack of 15 - $26.09
    $26.09
    Pack of 15
       $26.09
     

    Supplier Number: 2226995
    Specifications:
    Marketsource
    Marketsource
  • ±0.02
    Phosphate
    25 °C
    Pack of 250
    7
    Yellow
    Log in for availability
    Log in for availability
    Pack of 250
    Pack of 250 - $200.77
    $200.77
    Pack of 250
       $200.77
     

    Supplier Number: 2227064
    Specifications:
    Marketsource
    Marketsource
  • ±0.02
    Phosphate
    25 °C
    Pack of 50
    7
    Yellow
    Log in for availability
    Log in for availability
    Pack of 50
    Pack of 50 - $48.80
    $48.80
    Pack of 50
       $48.80
     

    Supplier Number: 2227066
    Specifications:
    Marketsource
    Marketsource
  • ±0.02
    Phosphate
    25 °C
    Pack of 15
    7
    Yellow
    Log in for availability
    Log in for availability
    Pack of 15
    Pack of 15 - $26.09
    $26.09
    Pack of 15
       $26.09
     

    Supplier Number: 2227095
    Specifications:
    Marketsource
    Marketsource
  • ±0.02
    Carbonate
    25 °C
    Pack of 250
    10.01
    Blue
    Log in for availability
    Log in for availability
    Pack of 250
    Pack of 250 - $200.77
    $200.77
    Pack of 250
       $200.77
     

    Supplier Number: 2227164
    Specifications:
    Marketsource
    Marketsource
  • ±0.02
    25 °C
    Pack of 20
    4.01 and 7.00
    Red/yellow
    Log in for availability
    Log in for availability
    Pack of 20
    Pack of 20 - $32.05
    $32.05
    Pack of 20
       $32.05
     

    Supplier Number: 2299264
    Specifications:
    Marketsource
    Marketsource