sensION™+ pH/mV/ion Meter, Bench
Catalog # 104087-434
Supplier: Hach
Specifications
- Description:pH/mV/ion kit, sensION™+ MM340
- Model:sensION™+ MM340
- Temperature Accuracy:≤0.2 °C
- Temperature Resolution:0.1 °C
- Connections:2 BNC, 2 ref, Pt1000
- Data Logging Capacity:Up to 400 readings per channel
- Display:Graphic, backlit liquid crystal, 128×64 dots
- ISE Calibration:2 - 5 programmable standards
- ISE Resolution:Programmable
- ISE Units:mol/L, M, mmol/L, mM, µmol/L, g/L, mg/L, ppm and %
- ISE Range:10⁻⁶ M - 10⁻¹ M
- Languages:English, german, spanish, french, italian and portuguese
- Outputs:RS232
- Temperature Compensation:Automatic with Pt 1000 temp. probe (ATC). or NTC 10 Kohm
- Weight:1.1 kg
- W×D×H:350×200×110 mm
- mV accuracy:≤0.2
- mV range:±2000
- mV resolution:0.1/1
- pH Accuracy:±0.002
- pH Calibration:1 - 3 point
- pH Range:−2.00...+16
- pH Resolution:0.1/0.001/0.01/0.1 (selectable)
- Cat. No.:104087-434
Specifications
About this item
This multi-parameter pH and ion-meter is perfect for research laboratories. It provides two measuring channels for pH/ORP/redox/ISE and option for two magnetic stirrers.
- Up to four ion selective calibration curves per channel; calibration with 2 to 5 standards
- Several calibration modes: with automatic recognition of 3 buffer groups, at a determined pH value and more
- Automatic buffer recognition: technical DIN 19267, DIN 19266 and up to 3 buffers chosen by the user
- Data logger storage of up to 330 measurements per channel
- RS232 C connectibility for printer or PC and connection for external PC, keyboard or barcode reader
Delivery: Supplied with integrated magnetic stirrer and probe holder, power supply with interchangeable plugs, pH 4, pH 7 and pH 10 calibration standard solutions (125 ml), KCl electrolyte solution (50 ml), 3 calibration beakers and printed documentation.