Molybdate Test Kit, MO-LR
104086-148
: Hach
:
- Description:Molybdate test kit, MO-LR
- Cat. No.:104086-148
- Range:0 - 3.0 mg/L Mo
Molybdates are used to inhibit corrosion in industrial and utility water systems. The reagents in the kit reacts with molybdate to form a blue color, which is measured with a color disc.
- Easy to use color disc
- Portable test kit for on-site measurement
- Effectively monitors the levels of molybdate corrosion inhibitors
: The kit includes two reagents, one color comparator box, one color disc, two stoppers, instructions, and a carrying case.