Orders LinkContactUsLinkComponent
Nitrification Inhibitors for BOD
Nitrification Inhibitors for BOD
Marketsource
Supplier:   Hach
Nitrification Inhibitors for BOD
Supplier:  Hach
Marketsource
Certificates
Nitrification Inhibitors for BOD
Nitrification Inhibitors for BOD
3 Options Available Below
Product Details & Documents
TCMP - 2-chloro-6 (trichloromethyl) pyridine (N-Serve), coated on an inert substrate.
Documents
Product Family Options
Product Information
  • Size
    Availability
    Price
  • 104086-382
    Nitrification inhibitor for BOD, Formula 2533™, TCMP, use 0.16 g in a 300 ml sample
    Nitrification inhibitor for BOD, Formula 2533™, TCMP, use 0.16 g in a 300 ml sample
    500 g
    Log in for availability
    Log in for availability
    Each
    Each - $447.23
    $447.23
    Each
       $447.23
    Minimum order quantity is 1
    Supplier Number: 253334
    Specifications:
    Marketsource
    Marketsource
  • 104086-384
    Nitrification inhibitor for BOD, contains 2-chloro-6 (trichloromethyl) pyridine (N-Serve), coated on an inert substrate, use two shots from cap dispenser in a 300 ml sample
    Nitrification inhibitor for BOD, contains 2-chloro-6 (trichloromethyl) pyridine (N-Serve), coated on an inert substrate, use two shots from cap dispenser in a 300 ml sample
    35 g
    Log in for availability
    Log in for availability
    Each
    Each - $55.78
    $55.78
    Each
       $55.78
    Minimum order quantity is 1
    Supplier Number: 253335
    Specifications:
    Marketsource
    Marketsource
Cap dispenser
Product Information
  • Size
    Availability
    Price
  • 104088-334
    Dispenser cap to fit 35 g bottle
    Dispenser cap to fit 35 g bottle
    Log in for availability
    Log in for availability
    Each
    Each - $49.83
    $49.83
    Each
       $49.83
    Minimum order quantity is 1
    Supplier Number: 45901
    Specifications:
    Marketsource
    Marketsource