To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Temperature Accuracy:≤0.2 °C
- Temperature Resolution:0.1 °C
- Compliance:IP 42
- Connections:BNC, banana, ATC type Pt 1000
- Display:Backlit graphical LCD, 128×64 pixel
- Temperature Compensation:Manual entry (via keypad) or with Pt 1000 temperature probe (ATC)
- Weight:1.1 kg
- W×D×H:350×201×110 mm
- mV accuracy:≤1 mV
- mV range:±1999
- mV resolution:1
- pH Accuracy:0.01
- pH Calibration:1 - 3 point
- pH Range:−2.00...+19.99
- pH Resolution:0.1/0.01 (selectable)
- Cat. No.:104087-406
Specifications
About this item
This simple pH meter fulfils all the requirements for routine measurements in industry and education. The all-in-one system with guided menu navigation makes general electrochemistry testing fast and simple. Each system is designed to be used in a wide variety of applications and comes complete with everything you need to start testing.
- Integrated magnetic stirrer for precise and repeatable measurements in accordance with GLP
- Labelled calibration flasks minimize the required buffer volume
- Calibration in 1, 2 or 3 points and mV calibration with Redox standard
- Alarm informs the user if the date of calibration is exceeded
- Large graphic display