Orders LinkContactUsLinkComponent
Test Kits, Drop Count Tests
Test Kits, Drop Count Tests
Marketsource
Supplier:   Hach
Test Kits, Drop Count Tests
Supplier:  Hach
Marketsource
Certificates
Test Kits, Drop Count Tests
Test Kits, Drop Count Tests
25 Options Available Below
Product Details & Documents
The drop count test is a simple, easy and cost-effective way to test for many parameters like chlorine, chloride, dissolved oxygen, carbon dioxide, sulphite, nitrogen, formaldehyde, and more.
  • Portable - ideal for use in field or in lab
  • Used as a quick reference or for confirmatory test with color indicator
  • Reagents supplied with the kit
Caution: Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
Product Family Options
Test kits
Product Information
  • Range
    Availability
    Price
  • 104085-424
    Carbon dioxide, model CA-23
    Carbon dioxide, model CA-23
    1.25 - 25. 2 - 40 and 5 - 100 mg/L
    Log in for availability
    Log in for availability
    Each
    Each - $112.42
    $112.42
    Each
       $112.42
    Minimum order quantity is 1
    Supplier Number: 143601
    Specifications:
    Marketsource
    Marketsource
  • 104085-508
    Hardness* (total+Ca), model HA-4P
    Hardness* (total+Ca), model HA-4P
    17 - 340 mg/L
    Log in for availability
    Log in for availability
    Each
    Each - $222.92
    $222.92
    Each
       $222.92
    Minimum order quantity is 1
    Supplier Number: 145700
    Specifications:
    Marketsource
    Marketsource
  • 104085-510
    Hardness* (total+Ca as CaCO₃+Mg), model HA-4P MG-L
    Hardness* (total+Ca as CaCO₃+Mg), model HA-4P MG-L
    20 - 400 mg/L
    Log in for availability
    Log in for availability
    Each
    Each - $222.92
    $222.92
    Each
       $222.92
    Minimum order quantity is 1
    Supplier Number: 145701
    Specifications:
    Marketsource
    Marketsource
  • 104086-084
    Hydrogen peroxide, model HYP-1
    Hydrogen peroxide, model HYP-1
    1 - 10 mg/L
    Log in for availability
    Log in for availability
    Each
    Each - $209.08
    $209.08
    Each
       $209.08
    Minimum order quantity is 1
    Supplier Number: 2291700
    Specifications:
    Marketsource
    Marketsource
  • 104086-096
    Ascorbic acid, model ASC-1
    Ascorbic acid, model ASC-1
    10 - 200 mg/L
    Log in for availability
    Log in for availability
    Each
    Each - $202.15
    $202.15
    Each
       $202.15
    Minimum order quantity is 1
    Supplier Number: 2308100
    Specifications:
    Marketsource
    Marketsource
  • 104086-576
    Hypochlorite, model CN-HR
    Hypochlorite, model CN-HR
    50000 - 150000 mg/L
    Log in for availability
    Log in for availability
    Each
    Each - $282.46
    $282.46
    Each
       $282.46
    Minimum order quantity is 1
    Supplier Number: 2687200
    Specifications:
    Marketsource
    Marketsource
  • 104085-482
    Total hardness, model HA-71A
    Total hardness, model HA-71A
    1 - 20 mg/L
    Log in for availability
    Log in for availability
    Each
    Each - $164.77
    $164.77
    Each
       $164.77
    Minimum order quantity is 1
    Supplier Number: 145201
    Specifications:
    Marketsource
    Marketsource
  • 104085-484
    Total hardness, model 5-B
    Total hardness, model 5-B
    17 - 510 mg/L
    Log in for availability
    Log in for availability
    Each
    Each - $49.22
    $49.22
    Each
       $49.22
    Minimum order quantity is 1
    Supplier Number: 145300
    Specifications:
    Marketsource
    Marketsource
  • 104085-488
    Total hardness, model 5-EP, 100 tests
    Total hardness, model 5-EP, 100 tests
    17 - 510 mg/L
    Log in for availability
    Log in for availability
    Each
    Each - $80.43
    $80.43
    Each
       $80.43
    Minimum order quantity is 1
    Supplier Number: 145400
    Specifications:
    Marketsource
    Marketsource
  • 104085-752
    Chloride, model CD-51
    Chloride, model CD-51
    500 - 10000 mg/L Cl-; 5000 - 100000 mg/L Cl-
    Log in for availability
    Log in for availability
    Each
    Each - $301.85
    $301.85
    Each
       $301.85
    Minimum order quantity is 1
    Supplier Number: 208601
    Specifications:
    Marketsource
    Marketsource
  • 104086-010
    Chlorine (Total), model CN-65
    Chlorine (Total), model CN-65
    1 - 20 mg/L
    Log in for availability
    Log in for availability
    Each
    Each - $217.38
    $217.38
    Each
       $217.38
    Minimum order quantity is 1
    Supplier Number: 225401
    Specifications:
    Marketsource
    Marketsource
  • 104085-456
    Chloride, model 8-P, use silver nitrate titrant
    Chloride, model 8-P, use silver nitrate titrant
    5 - 100 mg/L Cl-; 20 - 400 mg/L Cl-
    Log in for availability
    Log in for availability
    Each
    Each - $138.45
    $138.45
    Each
       $138.45
    Minimum order quantity is 1
    Supplier Number: 144001
    Specifications:
    Marketsource
    Marketsource
  • 104085-528
    Dissolved oxygen, model OX-2P
    Dissolved oxygen, model OX-2P
    1 - 20 mg/L O₂
    Log in for availability
    Log in for availability
    Each
    Each - $217.38
    $217.38
    Each
       $217.38
    Minimum order quantity is 1
    Supplier Number: 146900
    Specifications:
    Marketsource
    Marketsource
  • 104085-932
    Chromium, model CH-14
    Chromium, model CH-14
    50 - 1000 mg/L Cr(6+)
    Log in for availability
    Log in for availability
    Each
    Each - $177.23
    $177.23
    Each
       $177.23
    Minimum order quantity is 1
    Supplier Number: 222702
    Specifications:
    Marketsource
    Marketsource
  • 104086-276
    Alkalinity, model AL-AP
    Alkalinity, model AL-AP
    17 - 342 mg/L
    Log in for availability
    Log in for availability
    Each
    Each - $121.98
    $121.98
    Each
       $121.98
    Minimum order quantity is 1
    Supplier Number: 2444300
    Specifications:
    Marketsource
    Marketsource
  • 104088-116
    Sulphiite, model SU-5
    Sulphiite, model SU-5
    Log in for availability
    Log in for availability
    Each
    Each - $250.62
    $250.62
    Each
       $250.62
    Minimum order quantity is 1
    Supplier Number: 148002
    Specifications:
    Marketsource
    Marketsource
  • 104088-204
    Alkalinity, model AL-TA, 100 tests
    Alkalinity, model AL-TA, 100 tests
    385 - 8500 mg/L
    Log in for availability
    Log in for availability
    Each
    Each - $121.98
    $121.98
    Each
       $121.98
    Minimum order quantity is 1
    Supplier Number: 2314500
    Specifications:
    Marketsource
    Marketsource
  • 104088-170
    Formaldehyde, model FM-1
    Formaldehyde, model FM-1
    0.5 - 10%
    Log in for availability
    Log in for availability
    Each
    Each - $240.92
    $240.92
    Each
       $240.92
    Minimum order quantity is 1
    Supplier Number: 2183100
    Specifications:
    Marketsource
    Marketsource
Replacement reagent set
Product Information
  • Range
    Availability
    Price
  • 104086-262
    Carbon dioxide, reagent set for model CA-23, drop count titration, set includes phenolphthalein indicator solution and sodium hydroxide standard solution (0.01 N)
    Carbon dioxide, reagent set for model CA-23, drop count titration, set includes phenolphthalein indicator solution and sodium hydroxide standard solution (0.01 N)
    Log in for availability
    Log in for availability
    Each
    Each - $68.60
    $68.60
    Each
       $68.60
    Minimum order quantity is 1
    Supplier Number: 2437500
    Specifications:
    Marketsource
    Marketsource
  • 104086-264
    Chloride reagent set for 8-P, drop count titration, set includes chloride 2 indicator powder pillows and silver nitrate titrant (0.0493 N titrant)
    Chloride reagent set for 8-P, drop count titration, set includes chloride 2 indicator powder pillows and silver nitrate titrant (0.0493 N titrant)
    Log in for availability
    Log in for availability
    Each
    Each - $117.89
    $117.89
    Each
       $117.89
    Minimum order quantity is 1
    Supplier Number: 2437600
    Specifications:
    Marketsource
    Marketsource
  • 104086-266
    Chlorine (total) reagent set for CN-21P, includes sulfamic acid powder pillows, potassium iodide powder pillows and sodium thiosulfate standard solution (0.0246 N)
    Chlorine (total) reagent set for CN-21P, includes sulfamic acid powder pillows, potassium iodide powder pillows and sodium thiosulfate standard solution (0.0246 N)
    Log in for availability
    Log in for availability
    Each
    Each - $117.62
    $117.62
    Each
       $117.62
    Minimum order quantity is 1
    Supplier Number: 2437700
    Specifications:
    Marketsource
    Marketsource
  • 104086-268
    Hardness (total) reagent set for HA-71A, includes buffer solution (hardness 1), hardness 2 and hardness 3 test solutions
    Hardness (total) reagent set for HA-71A, includes buffer solution (hardness 1), hardness 2 and hardness 3 test solutions
    Log in for availability
    Log in for availability
    Each
    Each - $94.91
    $94.91
    Each
       $94.91
    Minimum order quantity is 1
    Supplier Number: 2437800
    Specifications:
    Marketsource
    Marketsource
  • 104086-272
    Sulphite reagent set for SU-5 kit, includes sulphamic acid powder pillows, sulphite 1 reagent powder pillows and sulphite 3 solution
    Sulphite reagent set for SU-5 kit, includes sulphamic acid powder pillows, sulphite 1 reagent powder pillows and sulphite 3 solution
    up to 200 mg/L SO₃
    Log in for availability
    Log in for availability
    Each
    Each - $150.92
    $150.92
    Each
       $150.92
    Minimum order quantity is 1
    Supplier Number: 2440000
    Specifications:
    Marketsource
    Marketsource
  • 104086-794
    Hardness 3 solution, 100 ml MDB
    Hardness 3 solution, 100 ml MDB
    Log in for availability
    Log in for availability
    Each
    Each - $21.86
    $21.86
    Each
       $21.86
    Minimum order quantity is 1
    Supplier Number: 42632
    Specifications:
    Marketsource
    Marketsource
Standards
Product Information
  • Range
    Availability
    Price
  • 104086-194
    Sodium thiosulfate standard solution, stabilised, 0.00246 N, 500 ml bottle
    Sodium thiosulfate standard solution, stabilised, 0.00246 N, 500 ml bottle
    Log in for availability
    Log in for availability
    Each
    Each - $67.08
    $67.08
    Each
       $67.08
    Minimum order quantity is 1
    Supplier Number: 2408549
    Specifications:
    Marketsource
    Marketsource