Orders LinkContactUsLinkComponent
Starch Indicator Solution, MDB
Starch Indicator Solution, MDB
Starch indicator solution, 100 ml
Catalog # 104086-774
Marketsource
Supplier:   Hach
Supplier #:  34932
Starch Indicator Solution, MDB
Catalog # 104086-774
Supplier:  Hach
Marketsource
CAS Number:  
Starch Indicator Solution, MDB
Starch Indicator Solution, MDB
Starch indicator solution, 100 ml
CAS Number:  
Price
$28.45
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Oxidation-reduction indicator for the analysis of BOD, total chlorine, chromium, cyanide, dissolved oxygen and sulfite.
  • Marked dropping bottle (MDB)
Documents
Specifications
Specifications
  • Description:
    Starch indicator solution
  • Size:
    100 ml
Specifications