Electrode Storage Bottles
Catalog # 104088-366
Supplier: Hach
Specifications
- Description:Storage bottle for electrodes, 12 ml
- Cat. No.:104088-366
Specifications
About this item
Electrode storage bottle is used to store pH electrodes.
- Rubber grommet in lid ensures tight seal with 12 mm electrodes