Hardness (Ca and Total Sequential) HR Reagent Set, Digital Titrator
Catalog # 104086-298
Supplier: Hach
CAS Number:
Specifications
- Chemical method:EDTA
- Description:No of tests: 100
- Cat. No.:104086-298
- Range:100 - 4000 mg/L CaCO₃
Specifications
About this item
For sequential determination of calcium and total hardness by EDTA digital titrator titration.
- Accurate results; when and where you need them
- Fast and flexible
- Versatile
- Economical
Hach method 8329. Range: 100 to 4000 mg/L CaCO₃. Set contains buffer solution (Hardness 1), CalVer® 2 calcium indicator powder pillows, ManVer® 2 hardness indicator powder pillows, potassium hydroxide solution (8 N), sulfuric acid (5.25 N) and 0.800 M EDTA digital titrator cartridge. Approximately 40 to 100 tests.
Ordering information: Hazardous items; items with this mark may be considered hazardous under some shipping conditions. If necessary, we will change your selected shipping method to accomodate these items.