NitraVer® 6 Nitrate Reagent powder Pillows
Catalog # MSPP-2107249
Supplier: HACH COMPANY MS
CAS Number:
Specifications
- Chemical method:Cadmium reduction
- Description:Nitrate NitraVer® 6, LR
- Cat. No.:MSPP-2107249
- Range:0.01 - 0.50 mg/L
- Sample Size:10 ml
- For unit:DR 6000; DR 3800; DR 2800; DR 2700; DR 1900; DR900
Specifications
About this item
These reagents are for the determination of low range nitrate by the NitraVer® 6 cadmium reduction method.