Orders LinkContactUsLinkComponent
TNTplus™ Alkalinity Kit (Total)
TNTplus™ Alkalinity Kit (Total)
Alkalinity, total, Colorimetric, DR 6000, DR 5000, DR 3900, DR3800, DR 2800, DR 1900, 25 - 400 mg/L CaCo₃, 25
Catalog # 104088-676
Marketsource
Supplier:   Hach
Supplier #:  TNT870
TNTplus™ Alkalinity Kit (Total)
Catalog # 104088-676
Supplier:  Hach
Marketsource
CAS Number:  
TNTplus™ Alkalinity Kit (Total)
TNTplus™ Alkalinity Kit (Total)
Alkalinity, total, Colorimetric, DR 6000, DR 5000, DR 3900, DR3800, DR 2800, DR 1900, 25 - 400 mg/L CaCo₃, 25
CAS Number:  
Price
$103.77
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This kit is used for the determination of alkalinity (total) by the colorimeteric method using TNTplus vials.
  • Easy and safe to handle
  • No reagent blank necessary
  • Automatic method detection
Specifications
Specifications
  • Chemical method:
    Colorimetric
  • Description:
    Alkalinity, total
  • Range:
    25 - 400 mg/L CaCo₃
  • No. of tests:
    25
  • For unit:
    DR 6000, DR 5000, DR 3900, DR3800, DR 2800, DR 1900