Sulphide Reagents 1 and 2
Supplier: Hach
Certificates
About this item
Used for sulphide determination by the methylene blue method.
- Range upto 800 µg/L
- Used with visual test kits
Frequently Bought Together
Used for sulphide determination by the methylene blue method.