Orders LinkContactUsLinkComponent
Detergents Reagent Solution
Detergents Reagent Solution
Detergent reagent solution in marked dropping bottle (MDB), 100 ml, Toluidine blue-O chloroform
Catalog # 104085-218
Marketsource
Supplier:   Hach
Supplier #:  105932
Detergents Reagent Solution
Catalog # 104085-218
Supplier:  Hach
Marketsource
CAS Number:  
Detergents Reagent Solution
Detergents Reagent Solution
Detergent reagent solution in marked dropping bottle (MDB), 100 ml, Toluidine blue-O chloroform
CAS Number:  
Price
$62.43
Log in for availability
Pack of 100
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Used for the determination of detergents (surfactants) by the crystal violet method. For use with storm water test kit.
Packaging: Reagent is packed in a marked dropping bottle.
Specifications
Specifications
  • Chemical method:
    Toluidine blue-O chloroform
  • Description:
    Detergent reagent solution in marked dropping bottle (MDB)
  • Size:
    100 ml