Dissolved Oxygen, Reagent Set
Catalog # 104086-136
Supplier: Hach
CAS Number:
Specifications
- Chemical method:Winkler
- Description:Dissolved oxygen reagent set, including burette
- Cat. No.:104086-136
- Range:1 - 10 mg/L O2
- Sample Size:300 ml
- No. of tests:100
Specifications
About this item
For determination of dissolved oxygen by the Winkler method (azide modification) and buret titration.