Visual Comparator Test Kits, Silica
Supplier: Hach
Certificates
About this item
Qualitative results using traditional color comparator disc methods.
- Portable - ideal for field testing
- Simple fast method
- Kits contain all required reagents and apparatus in a rugged carrying case for 100 tests
Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
Frequently Bought Together