ContactUsLinkComponent
Visual Color Kits, Alkalinity, AL-AP
Visual Color Kits, Alkalinity, AL-AP
Catalog # 104086-278
Supplier:  Hach
CAS Number:  
undefined
Visual Color Kits, Alkalinity, AL-AP
Catalog # 104086-278
Supplier:  Hach
Supplier Number:  2444301
CAS Number:  
Specifications
  • Description:
    Alkalinity (CaCO₃)
  • Cat. No.:
    104086-278
  • Range:
    5 - 100 mg/L
  • No. of tests:
    100
Specifications
About this item
Caused by the presence of carbonates, bicarbonates, hydroxides, and other dissolved salts, alkalinity is important to food and beverage processors, boiler operators, and aquatic biologists.
  • Kit utilises drop count titration method
  • Simple, economical titration method
Delivery: Rugged kit contains all required reagents, solutions, and glassware.