ContactUsLinkComponent
Visual Comparator Test Kits, Chlorine (Total), Model CN-21P
Visual Comparator Test Kits, Chlorine (Total), Model CN-21P
Catalog # 104086-280
Supplier:  Hach
CAS Number:  
undefined
Visual Comparator Test Kits, Chlorine (Total), Model CN-21P
Catalog # 104086-280
Supplier:  Hach
Supplier Number:  2444400
CAS Number:  
Specifications
  • Description:
    Chlorine (total), CN-21P
  • Cat. No.:
    104086-280
  • Range:
    10 - 200 mg/L
  • No. of tests:
    100
Specifications
About this item
Qualitative results using traditional drop count titrimetric methods.
  • Portable - ideal for field testing
  • Simple fast method
  • Kits contain all required reagents and apparatus in a rugged carrying case for 100 tests
Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.